Learn More
Cayman Chemical ANTIBODY GSK2656157 5mg
SDP
Supplier:  Cayman Chemical 173725 MG
Encompass_Preferred
A selective inhibitor of PERK (IC50 = 0.9 nM); blocks both stress-induced PERK autophosphorylation and eIF2α substrate phosphorylation and decreases levels of ATF4 and CHOP in multiple cell lines
Catalog No. 50-296-8232