Learn More
Cayman Chemical ANTIBODY GSK2801 5mg
SDP
Supplier:  Cayman Chemical 141205 MG
Encompass_Preferred
A selective, cell-permeable inhibitor of the BAZ2 family of bromodomain containing proteins (IC50s = 9-350 nM in a BAZ2B AlphaScreen; KDs = 0.26 and 0.14 µM, for BAZ2A and BAZ2B respectively)
Catalog No. 50-295-4404