Learn More
Cayman Chemical ANTIBODY GSK2982772 1mg
SDP
Supplier:  Cayman Chemical 292301 MG
Encompass_Preferred
A RIPK1 inhibitor (IC50s = 16 and 2,500 nM for the human and mouse enzymes, respectively); >10,000-fold selective for RIPK1 over a panel of 339 kinases at 10 µM; inhibits necrotic cell death induced by a combination of TNF-α and the caspase inhibitor QVD-OPh in U937 human monocytic and L929 murine fibrosarcoma cells (IC50s = 6.3 and 1,300 nM, respectively); decreases IL-1β and IL-6 levels in intestinal mucosa tissue isolated from patients with ulcerative colitis at 3-300 nM; increases survival in a mouse model of TNF-α-induced lethal shock at 3, 10, and 50 mg/kg
Catalog No. 50-293-7187