Learn More
Cayman Chemical ANTIBODY GSK2983559 1mg
SDP
Supplier:  Cayman Chemical 314581 MG
Encompass_Preferred
A RIPK2 inhibitor; inhibits RIPK2 by 65% in a kinase assay at 10 µM; inhibits VEGFR3 by greater than 90%, as well as 14 additional kinases by 60-89% in a panel of 344 kinases at 10 µM; reduces colonic damage in a mouse model of TNBS-induced colitis at 7.5 and 145 mg/kg twice per day; a prodrug
Catalog No. 50-299-1078