Learn More
Cayman Chemical ANTIBODY GSK3787 10mg
SDP
Supplier:  Cayman Chemical 1521910 MG
Encompass_Preferred
A selective, irreversible antagonist of PPARβ/δ (pIC50 = 6.6) that inhibits the expression of PPARβ/δ-regulated target genes, PDK4, CPT1a, and Angptl4
Catalog No. 50-297-2510