Learn More
Cayman Chemical ANTIBODY GSK5959 10mg
SDP
Supplier:  Cayman Chemical 1812310 MG
Encompass_Preferred
A potent, selective, and cell-permeable inhibitor of the BRPF1 bromodomain (IC50 = 80 nM); disrupts chromatin binding of BRPF1 in a cellular assay by blocking the interaction of BRPF1 with histone H3.3
Catalog No. 50-296-8230