Learn More
Cayman Chemical ANTIBODY GSK621 1mg
SDP
Supplier:  Cayman Chemical 191551 MG
Encompass_Preferred
An AMPK activator that reduces the proliferation of AML cell lines (IC50s = 13-30 μM), triggering autophagy via the eIF2α/ATF4 signaling pathway
Catalog No. 50-298-1180