Learn More
Cayman Chemical ANTIBODY GSK690693 10mg
SDP
Supplier:  Cayman Chemical 1689110 MG
Encompass_Preferred
An ATP-competitive, pan-Akt kinase inhibitor (IC50s = 2, 13, and 9 nM for Akt1, 2, and 3, respectively) that inhibits proliferation and induces apoptosis in various human tumor cells in vitro and in xenografts in immunocompromised mice
Catalog No. 50-296-6348