Learn More
Cayman Chemical ANTIBODY GSK923295 50mg
SDP
Supplier:  Cayman Chemical 1838950 MG
Encompass_Preferred
A potent inhibitor of CENP-E (Ki = 3.2 nM); causes mitotic cell cycle delay, leading to apoptosis in various cancer cell lines; induces apoptosis in cancer cells in mice bearing xenografts of COLO 205 cells
Catalog No. 50-296-6999