Learn More
Cayman Chemical ANTIBODY GTP 14564 5mg
SDP
Supplier:  Cayman Chemical 175435 MG
Encompass_Preferred
An inhibitor of class III receptor tyrosine kinases (IC50s = 0.3 µM for c-Fms, c-Kit, ITD-FLT3 and 1 µM for PDGFRβ); blocks the proliferation of leukemia cells stimulated with FLT3 ligand by preventing the activation of STAT5
Catalog No. 50-296-4885