Learn More
Cayman Chemical ANTIBODY GX-674 5mg
SDP
Supplier:  Cayman Chemical 396775 MG
Encompass_Preferred
A state-dependent inhibitor of Nav1.7; selectively inhibits Nav1.7 in HEK293 cells in a patch-clamp assay at -40 mV over -120 mV (IC50s = 0.1 and 240 nM, respectively); selective for Nav1.7 over Nav1.1, Nav1.3-1.5, and Nav1.8 but does inhibit Nav1.2 and, to a lesser extent, Nav1.6
Catalog No. 50-291-4735