Learn More
Cayman Chemical ANTIBODY GYY 4137 50mg
SDP
Supplier:  Cayman Chemical 1334550 MG
Encompass_Preferred
A water-soluble, slow-releasing H2S donor which demonstrates vasodilator and anti-hypertensive activity in hypertensive rats when given intravenously; also protects against endotoxic shock in rats, inhibiting pro-inflammatory signaling
Catalog No. 50-295-3786