Learn More
Cayman Chemical ANTIBODY H-151 50mg
SDP
Supplier:  Cayman Chemical 2585750 MG
Encompass_Preferred
A STING inhibitor; forms an adduct with mouse STING in serum and acts in an irreversible manner; inhibits the type I IFN response in vitro; reduces phosphorylation of TBK1 and palmitoylation of hsSTING; reduces CMA-induced increases in mouse serum levels of IFN-β and IL-6
Catalog No. 50-293-4338