Learn More
Cayman Chemical ANTIBODY HAPC-Chol 25mg
SDP
Supplier:  Cayman Chemical 2658425 MG
Encompass_Preferred
A cationic cholesterol; has been used for siRNA delivery and gene silencing in MCF-7 cells in a luciferase assay without affecting cell viability; has been used to deliver siRNA into mice via intravenous injection, resulting in HAPC-chol accumulation in the lungs
Catalog No. 50-293-4428