Learn More
Cayman Chemical ANTIBODY Harmol 1mg
SDP
Supplier:  Cayman Chemical 338441 MG
Encompass_Preferred
An analytical reference standard categorized as a β-carboline alkaloid; has been found in Psilocybe mushrooms; intended for research and forensic applications
Catalog No. 50-294-8025