Learn More
Cayman Chemical ANTIBODY HCV-IN-31 25mg
SDP
Supplier:  Cayman Chemical 3677825 MG
Encompass_Preferred
An antiviral nucleoside; inhibits HCV replication in an HCV replicon assay (EC50 = 15.7 µM)
Catalog No. 50-291-3330