Learn More
Cayman Chemical ANTIBODY HDAC8-IN-1 5mg
SDP
Supplier:  Cayman Chemical 288325 MG
Encompass_Preferred
An HDAC8 inhibitor (IC50 = 27.2 nM); selective for HDAC8 over HDAC1-3, -4, -6, -10, and -11 (IC50s = ≥3,000 nM for all); cytotoxic to A549, H1299, and CL1-5 lung cancer cells (IC50s = 7.9, 7.2, and 7 μM, respectively)
Catalog No. 50-294-0048