Learn More
Cayman Chemical ANTIBODY HesperadIn 10mg
SDP
Supplier:  Cayman Chemical 2419910 MG
Encompass_Preferred
A multi-kinase inhibitor; inhibits human Aurora kinase B (IC50 = 250 nM) and the T. brucei homolog Aurora kinase-1 (IC50 = 40 nM); inhibits AMPK, LCK, MKK1, MAPKAP-K1, CHK1, and PHK in a panel of 25 kinases at 1 µM; inhibits MEKK2 in ATPase and transphosphorylation assays (IC50s = 60 and 34 nM, respectively); induces polyploidy and defects in cytokinesis and spindle assembly as well as inhibits proliferation of HeLa cells and overrides paclitaxel- and monastrol-induced mitotic arrest from 50-100 nM; induces toxicity in HepG2 cells (TC50 = 50 = 0.22-2.21 µM); inhibits the growth of T. brucei, L. major promastigotes and amasigotes, and P. falciparum (EC50s = 0.01-2.37 µM)
Catalog No. 50-293-3339