Learn More
Cayman Chemical ANTIBODY HesperIdIn 25g
SDP
Supplier:  Cayman Chemical 1864625 G
Encompass_Preferred
A flavanone rutinoside found in citrus peels with antioxidant, anticancer, and estrogenic effects in in vitro and in vivo research models; increases the cytotoxicity of doxorubicin on MCF-7 and HeLa cancer cells; decreases serum and hepatic lipid concentrations and reduces osteoporosis in ovariectomized rats
Catalog No. 50-298-0637