Learn More
Cayman Chemical ANTIBODY Hexestrol 5g
SDP
Supplier:  Cayman Chemical 290035 G
Encompass_Preferred
A synthetic non-steroidal estrogen that binds to ERα and ERβ (Kis = 0.06 and 0.06 nM for the human and rat receptors, respectively); inhibits rat liver microsomal and ox brain phospholipid liposomal lipid peroxidation (IC50s = 1.5 and 2.75 μM, respectively); inhibits porcine microtubule assembly and induces disassembly of preformed microtubules at 50 and 100 μM, respectively, in a cell-free assay; induces kidney tumors in male Syrian hamsters at 25 mg/animal
Catalog No. 50-294-1859