Learn More
Cayman Chemical ANTIBODY HIstamIn 25g
SDP
Supplier:  Cayman Chemical 3382825 G
Encompass_Preferred
A biogenic amine; activates histamine H1-4 receptors to regulate allergic and inflammatory responses, gastric acid secretion, arousal, cognition, and immune cell chemotaxis
Catalog No. 50-294-7137