Learn More
Cayman Chemical ANTIBODY HM43239 10mg
SDP
Supplier:  Cayman Chemical 3773710 MG
Encompass_Preferred
An FLT3 inhibitor (IC50s = 1.1 and 1.8 nM for the WT and ITD mutant, respectively); inhibits VEGFR2 and Syk by 100 and 95%, respectively, at 100 nM; inhibits the growth of MOLM-13 and MV4-11 leukemia cells (GI50s = 5.2 and 1.3 nM, respectively)
Catalog No. 50-291-7637