Learn More
Cayman Chemical ANTIBODY HMN-214 25mg
SDP
Supplier:  Cayman Chemical 2621025 MG
Encompass_Preferred
A prodrug form of the PLK inhibitor HMN-176; decreases MDR1 expression in AB-A.1 cells and in tumors isolated from mice bearing multidrug resistant KB-A.1. xenografts; reduces tumor volume in PC3, WiDr, and A549 mouse xenograft models at 20 mg/kg per day; does not induce nerve toxicity in rabbit sciatic nerves in vivo at 30 mg/kg per day
Catalog No. 50-293-5077