Learn More
Cayman Chemical ANTIBODY HPOB 1mg
SDP
Supplier:  Cayman Chemical 150661 MG
Encompass_Preferred
A potent, selective inhibitor of HDAC6 (IC50 = 56 nM); induces acetylation of α-tubulin but not histones; enhances the cytotoxicity of the broad spectrum HDAC inhibitor SAHA against cancer cells in nude mice carrying an androgen-dependent CWR22 human prostate cancer xenograft
Catalog No. 50-297-1630