Learn More
Cayman Chemical ANTIBODY HS-1371 10mg
SDP
Supplier:  Cayman Chemical 3103310 MG
Encompass_Preferred
A RIPK3 inhibitor (IC50 = 20.8 nM); inhibits basal RIPK3 autophosphorylation and TNF-α- or TRAIL-induced necroptosis in HT-29 cells at 5 µM; inhibits TNF-α Smac mimetic- and Z-VAD-induced necroptosis in HeLa and NCI-H2009 cancer cells ectopically expressing RIPK3
Catalog No. 50-298-9503