Learn More
Cayman Chemical ANTIBODY HUP-55 5mg
SDP
Supplier:  Cayman Chemical 391175 MG
Encompass_Preferred
A prolyl endopeptidase inhibitor (IC50 = 5 nM); reduces α-synuclein dimerization in Neuro2a cells, induces autophagy in HEK293 cells, and decreases hydrogen peroxide-induced increases in ROS production in SH-SY5Y cells at 10 µM; decreases ipsilateral paw use and striatal oligomeric α-synuclein levels in an AAV-αSyn mouse model of Parkinson’s disease at 10 mg/kg
Catalog No. 50-291-7258