Learn More
Cayman Chemical ANTIBODY HY-17542 10mg
SDP
Supplier:  Cayman Chemical 3335410 MG
Encompass_Preferred
A SARS-CoV PLpro inhibitor (IC50 = 2.6 µM); increases the viability of SARS-CoV-infected Vero E6 cells (EC50 = 13.1 µM)
Catalog No. 50-294-7236