Learn More
Cayman Chemical ANTIBODY IbIglustat 50mg
SDP
Supplier:  Cayman Chemical 3287850 MG
Encompass_Preferred
A GlcCer synthase inhibitor; reduces Gb3 and lyso-Gb3 accumulation in a variety of tissues in the Gla-/- mouse model of Fabry disease; reduces the latency to paw withdrawal in the hot plate test in the same model; reduces gliosis, delays the onset of ataxia, and increases lifespan in the transgenic 4L;C* mouse model of neuronopathic Gaucher disease at 0.03% w/w in the diet
Catalog No. 50-294-8743