Learn More
Cayman Chemical ANTIBODY IcarItIn 5mg
SDP
Supplier:  Cayman Chemical 202365 MG
Encompass_Preferred
A flavonoid that inhibits FASN, reducing IGF-1-induced activation of STAT3 in several melanoma cell lines
Catalog No. 50-298-1167