Learn More
Cayman Chemical ANTIBODY ICG-001 1mg
SDP
Supplier:  Cayman Chemical 162571 MG
Encompass_Preferred
A small molecule inhibitor of β-catenin/cyclic AMP responsive element binding protein (CREB) binding protein (CBP)-mediated transcription (IC50 = 3 µM); selectively induces apoptosis in transformed colon cells but not in normal cells and prevents the growth of colon carcinoma cells at 25 µM in vitro
Catalog No. 50-297-2903