Learn More
Cayman Chemical ANTIBODY IDO-IN-1 1mg
SDP
Supplier:  Cayman Chemical 194021 MG
Encompass_Preferred
A potent IDO inhibitor (IC50s= 59 and 12 nM for human recombinant and HeLa cell IDO, respectively); inactive against TOD (IC50 > 10 µM).
Catalog No. 50-298-2779