Learn More
Cayman Chemical ANTIBODY IEM 1460 50mg
SDP
Supplier:  Cayman Chemical 1562350 MG
Encompass_Preferred
An adamantine derivative that selectively blocks Ca2+-permeable glutamate receptor channels (IC50 = 10 μM-0.1 mM)
Catalog No. 50-297-3211