Learn More
Cayman Chemical ANTIBODY IfosfamIde 1g
SDP
Supplier:  Cayman Chemical 175621 G
Encompass_Preferred
A nitrogen mustard alkylating agent used as a chemotherapeutic agent against advanced transitional cell carcinoma; serves as a prodrug that is metabolized to its active DNA alkylating form, isophosphoramide mustard
Catalog No. 50-298-0084