Learn More
Cayman Chemical ANTIBODY IMR-1 25mg
SDP
Supplier:  Cayman Chemical 2792025 MG
Encompass_Preferred
An inhibitor of Maml1 recruitment to chromatin; binds to the intracellular domain of Notch (KD = 11 µM) and prevents transcription of Notch target genes by inhibiting the recruitment of Maml1 to chromatin; inhibits colony formation in Notch-dependent cell lines but not Notch-independent cell lines at 15 and 45 µM; decreases the expression of Notch target genes and reduces tumor growth in a patient-derived esophageal adenocarcinoma mouse xenograft model at 15 mg/kg
Catalog No. 50-293-9735