Learn More
Cayman Chemical ANTIBODY INK1117 25mg
SDP
Supplier:  Cayman Chemical 1951425 MG
Encompass_Preferred
An inhibitor of PI3Kα that is selective for p110α in vitro (IC50 = 15 nM for p110α vs. >1 µM for other isoforms, as well as for mTOR) and in cells when used at 1 µM
Catalog No. 50-298-0937