Learn More
Cayman Chemical ANTIBODY IP7e 5mg
SDP
Supplier:  Cayman Chemical 297265 MG
Encompass_Preferred
An activator of Nurr1 signaling (EC50 = 3.9 nM in a reporter assay); prevents spinal cord axonal loss and demyelination and decreases spinal cord macrophage and T cell infiltration in a mouse model of EAE at 10 mg/kg
Catalog No. 50-298-9173