Learn More
Cayman Chemical ANTIBODY IPG-2 AM 500ug
SDP
Supplier:  Cayman Chemical 35540500 UG
Encompass_Preferred
A cell-permeable fluorescent potassium indicator; binds to potassium (Kd = 18 mM); ex/em = 525/545 nm, respectively; has been used to kinetically monitor potassium levels in isolated human platelets via flow cytometry
Catalog No. 50-291-0096