Learn More
Cayman Chemical ANTIBODY IPPD 100g
SDP
Supplier:  Cayman Chemical 40725100 G
Encompass_Preferred
A substituted p-phenylenediamine; reduces SOD and CAT activity in zebrafish larvae; reduces heart rate and induces cardiac malformations in zebrafish embryos at 300 UG/L; reduces the swimming speed of zebrafish larva at 2 and 300 UG/L but not 20 UG/L; has been found in rubber boots, particulate matter 2.5 (PM2.5), wastewater influent, effluent, and biosolids, and human milk
Catalog No. 50-297-8584