Learn More
Cayman Chemical ANTIBODY ISCK03 25mg
SDP
Supplier:  Cayman Chemical 1578125 MG
Encompass_Preferred
A cell-permeable inhibitor of SCF-mediated c-kit activation, completely blocking phosphorylation of c-kit in human melanoma cells at a concentration between 1 and 5 µM; prevents SCF-mediated downstream phosphorylation of p44/p42 ERK
Catalog No. 50-296-9501