Learn More
Cayman Chemical ANTIBODY IsoIndIgo 500mg
SDP
Supplier:  Cayman Chemical 33208500 MG
Encompass_Preferred
A building block; has been used in the synthesis of polymers for use in organic photovoltaics
Catalog No. 50-294-8498