Learn More
Cayman Chemical ANTIBODY IsonIazId 100g
SDP
Supplier:  Cayman Chemical 20378100 G
Encompass_Preferred
An antibiotic that acts as a prodrug that is converted to isonicotinic acyl-NADH complex, which inhibits mycolic acid biosynthesis; effective against several species of Mycobacterium, including M. tuberculosis
Catalog No. 50-298-0739