Learn More
Cayman Chemical ANTIBODY ISX-9 25mg
SDP
Supplier:  Cayman Chemical 1616525 MG
Encompass_Preferred
A small molecule inducer of adult neural stem cell differentiation; 2.5-20 µM triggers neurogenesis and blocks gliogenesis in HCN cells through a neurotransmitter-mediated calcium signal and myocyte-enhancer factor 2-dependent gene expression pathway
Catalog No. 50-297-1676