Learn More
Cayman Chemical ANTIBODY ITH15004 5mg
SDP
Supplier:  Cayman Chemical 336675 MG
Encompass_Preferred
A P2X7 receptor antagonist (IC50 = 9 µM); inhibits ATP-induced currents in X. laevis oocytes expressing the human P2X7 receptor at 100 µM; decreases IL-1β release from LPS-primed, ATP-stimulated isolated mouse peritoneal macrophages at 1 µM; has high permeability in a PAMPA
Catalog No. 50-294-6736