Learn More
Cayman Chemical ANTIBODY JaceosIdIn 1mg
SDP
Supplier:  Cayman Chemical 197581 MG
Encompass_Preferred
A natural methylated trihydroxyflavone; has antioxidative actions on cells in culture; induces cell cycle arrest and apoptosis in cancer cells; blocks cell signaling related to inflammation
Catalog No. 50-298-2907