Learn More
Cayman Chemical ANTIBODY JSH-23 25mg
SDP
Supplier:  Cayman Chemical 1503625 MG
Encompass_Preferred
An inhibitor of NF-κB, blocking its translocation into the nucleus (IC50 = 7.1 µM); does not affect degradation of IκB; interferes with NF-κB-mediated gene expression and apoptosis in RAW 264.7 cells treated with LPS
Catalog No. 50-296-9907