Learn More
Cayman Chemical ANTIBODY JT010 10mg
SDP
Supplier:  Cayman Chemical 2973210 MG
Encompass_Preferred
A TRPA1 agonist (EC50 = 0.65 nM in a calcium influx assay); selective for TRPA1 over TRPV1, TRPV3, and TRPV4, TRPC5, TRPM2, and TRPM8 channels at 1 µM
Catalog No. 50-298-9521