Learn More
Cayman Chemical ANTIBODY JWH 022 5mg
SDP
Supplier:  Cayman Chemical 90010565 MG
Encompass_Preferred
A cannabimimetic indole that is structurally related to JWH 018, a mildly selective agonist of the peripheral cannabinoid (CB2) receptor
Catalog No. 50-289-4589