Learn More
Cayman Chemical ANTIBODY JYL1421 1mg
SDP
Supplier:  Cayman Chemical 204581 MG
Encompass_Preferred
A TRPV1 antagonist; inhibits capsaicin-induced calcium uptake in CHO cells expressing rat TRPV1 (EC50 = 9.2 nM); inhibits capsaicin-induced release of somatostatin, substance P, and CGRP from isolated rat trachea (IC50s = 227-491 nM); inhibits capsaicin-induced hypothermia and hypotension in rats at 2 and 0.4 mg/kg, respectively; reduces the number of wiping movements induced by ocular administration of capsaicin in rats at 2 mg/kg
Catalog No. 50-298-3674