Learn More
Cayman Chemical ANTIBODY JZP 430 25mg
SDP
Supplier:  Cayman Chemical 3538325 MG
Encompass_Preferred
An ABHD6 inhibitor (IC50 = 0.044 for the human enzyme); selective for ABHD6 over FAAH and MAGL at 2.5 µM, ABHD12 at 1 µM, as well as LAL and CB1 and CB2 receptors at 10 µM
Catalog No. 50-291-3641