Learn More
Cayman Chemical ANTIBODY KC01 500ug
SDP
Supplier:  Cayman Chemical 17364500 UG
Encompass_Preferred
A covalent inhibitor of ABHD16A (IC50s = 90 and 520 nM for human and mouse, respectively); depletes lyso-PS from various human cancer cell lines, a lymphoblast cell line, and mouse brain membrane lysates and decreases LPS-induced cytokine production in macrophages
Catalog No. 50-297-9690